Assay Details
| Gene Symbol: LOC4329597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771824.2 | XP_015627310.1 | 3 - 4 | 2468 | 67 | |
| XM_015771825.2 | XP_015627311.1 | 3 - 4 | 2468 | 67 | ||
| XM_015771826.2 | XP_015627312.1 | 4 - 5 | 2772 | 67 | ||
| XM_015771827.2 | XP_015627313.1 | 4 - 5 | 2834 | 67 | ||
| XM_015771828.2 | XP_015627314.1 | 4 - 5 | 2766 | 67 | ||
| XM_015771829.2 | XP_015627315.1 | 4 - 5 | 2828 | 67 | ||
| XM_015771831.2 | XP_015627317.1 | 4 - 5 | 2758 | 67 | ||
| XM_015771832.2 | XP_015627318.1 | 4 - 5 | 2820 | 67 | ||
| GenBank mRNA | AK066798.1 | - | 3 - 4 | 2465 | 67 | |
More Information
Set Membership: |
|

LOC4329597
Genomic Map