Assay Details
| Gene Symbol: LOC4329732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769227.2 | XP_015624713.1 | 1 | 1426 | 108 | |
| GenBank mRNA | AK109591.1 | - | 1 | 1146 | 108 | |
More Information
Set Membership: |
|

LOC4329732
Genomic Map