Assay Details
| Gene Symbol: LOC4330582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769708.2 | XP_015625194.1 | 12 - 13 | 1623 | 81 | |
| GenBank mRNA | AK058309.1 | - | 11 - 12 | 1075 | 81 | |
| AK103602.1 | - | 12 - 13 | 1629 | 81 | ||
More Information
Set Membership: |
|

LOC4330582
Genomic Map