Assay Details
| Gene Symbol: LOC4330582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769708.2 | XP_015625194.1 | 5 - 6 | 785 | 61 | |
| GenBank mRNA | AK058309.1 | - | 4 - 5 | 237 | 61 | |
| AK103602.1 | - | 5 - 6 | 791 | 61 | ||
More Information
Set Membership: |
|

LOC4330582
Genomic Map