Assay Details
Gene Symbol: LOC4331680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776503.2 | XP_015631989.1 | 4 - 5 | 974 | 102 | |
GenBank mRNA | AK242248.1 | - | 4 - 5 | 960 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
