Assay Details
Gene Symbol: LOC4331680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776503.2 | XP_015631989.1 | 5 - 6 | 1370 | 88 | |
GenBank mRNA | AK242248.1 | - | 5 - 6 | 1355 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
