Assay Details
Gene Symbol: LOC4331680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776503.2 | XP_015631989.1 | 7 - 8 | 1572 | 104 | |
GenBank mRNA | AK242248.1 | - | 7 - 8 | 1557 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
