Assay Details
| Gene Symbol: LOC4331842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776790.2 | XP_015632276.1 | 7 - 8 | 632 | 60 | |
| GenBank mRNA | AK073293.1 | - | 7 - 8 | 617 | 60 | |
| AK120235.1 | - | 7 - 8 | 617 | 60 | ||
More Information
Set Membership: |
|

LOC4331842
Genomic Map