Assay Details
Gene Symbol: LOC4331881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772803.2 | XP_015628289.1 | 2 - 3 | 490 | 68 | |
GenBank mRNA | AK068057.1 | - | 2 - 3 | 486 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
