Assay Details
| Gene Symbol: LOC4332556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777733.2 | XP_015633219.1 | 6 - 7 | 4718 | 98 | |
| GenBank mRNA | AK243032.1 | - | 6 - 7 | 3948 | 98 | |
| AK289171.1 | - | 6 - 7 | 3948 | 98 | ||
More Information
Set Membership: |
|

LOC4332556
Genomic Map