Assay Details
| Gene Symbol: LOC4332645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775530.2 | XP_015631016.1 | 6 - 7 | 2171 | 153 | |
| XM_015775531.2 | XP_015631017.1 | 5 - 6 | 1761 | 153 | ||
| GenBank mRNA | AK064890.1 | - | 5 - 6 | 2524 | 153 | |
More Information
Set Membership: |
|

LOC4332645
Genomic Map