Assay Details
Gene Symbol: LOC4332650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777462.2 | XP_015632948.1 | 1 - 2 | 766 | 79 | |
GenBank mRNA | AK103334.1 | - | 1 - 2 | 764 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
