Assay Details
Gene Symbol: LOC4332650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777462.2 | XP_015632948.1 | 5 - 6 | 1694 | 63 | |
GenBank mRNA | AK103334.1 | - | 5 - 6 | 1692 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
