Assay Details
Gene Symbol: LOC4332695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015773510.2 | XP_015628996.1 | 1 - 2 | 744 | 152 | |
GenBank mRNA | AK108122.1 | - | 1 - 2 | 450 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
