Assay Details
| Gene Symbol: LOC4332771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774981.1 | XP_015630467.1 | 3 - 4 | 441 | 73 | |
| GenBank mRNA | AF119232.1 | - | 2 - 3 | 212 | 73 | |
| AK070859.1 | - | 3 - 4 | 424 | 73 | ||
More Information
Set Membership: |
|

LOC4332771
Genomic Map