Assay Details
| Gene Symbol: LOC4332846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777123.2 | XP_015632609.1 | 1 - 2 | 145 | 81 | |
| GenBank mRNA | AK061662.1 | - | 1 - 2 | 91 | 81 | |
| AK120692.1 | - | 1 - 2 | 83 | 81 | ||
| BT129038.1 | - | 1 - 2 | 83 | 81 | ||
| BT130719.1 | - | 1 - 2 | 80 | 81 | ||
| D00999.1 | - | 1 - 2 | 61 | 81 | ||
| L36320.1 | - | 1 - 2 | 75 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777123.2 transcripts.
Set Membership: |
|

LOC4332846
Genomic Map