Assay Details
| Gene Symbol: LOC4332846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777122.2 | XP_015632608.1 | 3 - 4 | 281 | 89 | |
| XM_015777123.2 | XP_015632609.1 | 3 - 4 | 337 | 89 | ||
| GenBank mRNA | AK061662.1 | - | 3 - 4 | 283 | 89 | |
| AK120692.1 | - | 3 - 4 | 275 | 89 | ||
| BT129038.1 | - | 3 - 4 | 275 | 89 | ||
| BT130719.1 | - | 3 - 4 | 272 | 89 | ||
| D00999.1 | - | 3 - 4 | 253 | 89 | ||
| L36320.1 | - | 3 - 4 | 89 | |||
More Information
Set Membership: |
|

LOC4332846
Genomic Map