Assay Details
| Gene Symbol: LOC4333066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777324.2 | XP_015632810.1 | 17 - 18 | 1457 | 151 | |
| XM_015777325.2 | XP_015632811.1 | 17 - 18 | 1457 | 151 | ||
| XM_015777326.2 | XP_015632812.1 | 17 - 18 | 1457 | 151 | ||
| GenBank mRNA | AK073203.1 | - | 17 - 18 | 1444 | 151 | |
More Information
Set Membership: |
|

LOC4333066
Genomic Map