Assay Details
| Gene Symbol: LOC4333066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777324.2 | XP_015632810.1 | 36 - 37 | 3418 | 136 | |
| GenBank mRNA | AK073203.1 | - | 36 - 37 | 3405 | 136 | |
More Information
Set Membership: |
|

LOC4333066
Genomic Map