Assay Details
| Gene Symbol: LOC4333841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775411.1 | XP_015630897.1 | 5 - 6 | 743 | 62 | |
| GenBank mRNA | AK059920.1 | - | 1 - 2 | 71 | 62 | |
| AK100165.1 | - | 5 - 6 | 630 | 62 | ||
| AK103027.1 | - | 5 - 6 | 626 | 62 | ||
| AK105026.2 | - | 5 - 6 | 627 | 62 | ||
| KF214527.1 | - | 5 - 6 | 620 | 62 | ||
| KM982771.1 | - | 5 - 6 | 602 | 62 | ||
| U28047.3 | - | 5 - 6 | 635 | 62 | ||
More Information
Set Membership: |
|

LOC4333841
Genomic Map