Assay Details
| Gene Symbol: LOC107278587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK069094.1 | - | 15 - 16 | 3410 | 112 | |
| Gene Symbol: LOC112936008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777255.2 | XP_015632741.1 | 5 - 6 | 1408 | 112 | |
| GenBank mRNA | AK102972.1 | - | 5 - 6 | 654 | 112 | |
More Information
Set Membership: |
|

LOC107278587
Genomic Map