Assay Details
| Gene Symbol: LOC4333884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777861.2 | XP_015633347.1 | 9 - 10 | 1616 | 121 | |
| XM_015777862.2 | XP_015633348.1 | 10 - 11 | 1247 | 121 | ||
| GenBank mRNA | AK060694.1 | - | 9 - 10 | 711 | 121 | |
| AK102350.1 | - | 10 - 11 | 1260 | 121 | ||
More Information
Set Membership: |
|

LOC4333884
Genomic Map