Assay Details
| Gene Symbol: LOC4333884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777861.2 | XP_015633347.1 | 1 - 2 | 932 | 91 | |
| XM_015777862.2 | XP_015633348.1 | 2 - 3 | 563 | 91 | ||
| GenBank mRNA | AK102350.1 | - | 2 - 3 | 576 | 91 | |
More Information
Set Membership: |
|

LOC4333884
Genomic Map