Assay Details
Gene Symbol: LOC4333953 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772784.2 | XP_015628270.1 | 4 - 5 | 580 | 60 | |
XM_015772785.2 | XP_015628271.1 | 4 - 5 | 578 | 60 | ||
GenBank mRNA | AK068261.1 | - | 4 - 5 | 495 | 60 | |
BT129118.1 | - | 2 - 3 | 194 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
