Assay Details
| Gene Symbol: LOC4333987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772747.2 | XP_015628233.1 | 1 - 2 | 392 | 79 | |
| GenBank mRNA | AK067045.1 | - | 1 - 2 | 383 | 79 | |
| AK099420.1 | - | 1 - 2 | 383 | 79 | ||
More Information
Set Membership: |
|

LOC4333987
Genomic Map