Assay Details
| Gene Symbol: LOC4333987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772747.2 | XP_015628233.1 | 3 - 4 | 553 | 64 | |
| GenBank mRNA | AK059020.1 | - | 1 - 2 | 64 | 64 | |
| AK067045.1 | - | 3 - 4 | 544 | 64 | ||
| AK099420.1 | - | 3 - 4 | 544 | 64 | ||
More Information
Set Membership: |
|

LOC4333987
Genomic Map