Assay Details
Gene Symbol: LOC4333987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772747.2 | XP_015628233.1 | 5 - 6 | 660 | 87 | |
GenBank mRNA | AK059020.1 | - | 3 - 4 | 171 | 87 | |
AK099420.1 | - | 5 - 6 | 651 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
