Assay Details
Gene Symbol: LOC4334140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775547.2 | XP_015631033.1 | 4 - 5 | 804 | 95 | |
XM_015775548.2 | XP_015631034.1 | 4 - 5 | 801 | 95 | ||
GenBank mRNA | AY332478.1 | - | 4 - 5 | 516 | 95 | |
AY551916.1 | - | 4 - 5 | 529 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
