Assay Details
Gene Symbol: LOC4334140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775548.2 | XP_015631034.1 | 6 - 7 | 888 | 68 | |
GenBank mRNA | AY332478.1 | - | 6 - 7 | 603 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
