Assay Details
Gene Symbol: LOC4334140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775547.2 | XP_015631033.1 | 7 - 8 | 1031 | 103 | |
XM_015775548.2 | XP_015631034.1 | 7 - 8 | 1007 | 103 | ||
GenBank mRNA | AY332478.1 | - | 7 - 8 | 722 | 103 | |
AY551916.1 | - | 7 - 8 | 756 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
