Assay Details
| Gene Symbol: LOC4329993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770112.2 | XP_015625598.1 | 6 - 7 | 1154 | 98 | |
| GenBank mRNA | AK106578.1 | - | 6 - 7 | 885 | 98 | |
| FJ008126.1 | - | 6 - 7 | 890 | 98 | ||
More Information
Set Membership: |
|

LOC4329993
Genomic Map