Assay Details
| Gene Symbol: LOC4330285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769956.2 | XP_015625442.1 | 1 - 2 | 354 | 85 | |
| GenBank mRNA | AK059611.1 | - | 1 - 2 | 323 | 85 | |
| AK099179.1 | - | 1 - 2 | 321 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769956.2 transcripts.
Set Membership: |
|

LOC4330285
Genomic Map