Assay Details
| Gene Symbol: LOC4331104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771492.2 | XP_015626978.1 | 8 - 9 | 1569 | 71 | |
| GenBank mRNA | AK065919.1 | - | 8 - 9 | 1535 | 71 | |
| AK119450.1 | - | 8 - 9 | 1536 | 71 | ||
More Information
Set Membership: |
|

LOC4331104
Genomic Map