Assay Details
Gene Symbol: LOC4332058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774590.2 | XP_015630076.1 | 1 - 2 | 216 | 61 | |
GenBank mRNA | AB106553.1 | - | 1 - 2 | 168 | 61 | |
AY494713.1 | - | 1 - 2 | 120 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
