Assay Details
Gene Symbol: LOC4332058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774589.2 | XP_015630075.1 | 2 - 3 | 358 | 68 | |
XM_015774590.2 | XP_015630076.1 | 2 - 3 | 352 | 68 | ||
GenBank mRNA | AB106553.1 | - | 2 - 3 | 304 | 68 | |
AY494713.1 | - | 2 - 3 | 256 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
