Assay Details
Gene Symbol: LOC4332058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774589.2 | XP_015630075.1 | 5 - 6 | 586 | 74 | |
XM_015774590.2 | XP_015630076.1 | 5 - 6 | 580 | 74 | ||
GenBank mRNA | AB106553.1 | - | 5 - 6 | 532 | 74 | |
AY494713.1 | - | 5 - 6 | 484 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
