Assay Details
Gene Symbol: LOC9270576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772424.1 | XP_015627910.1 | 6 - 7 | 564 | 87 | |
GenBank mRNA | AK072434.1 | - | 4 - 5 | 1973 | 87 | |
BT132393.1 | - | 6 - 7 | 504 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
