Assay Details
| Gene Symbol: LOC9270576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772424.1 | XP_015627910.1 | 8 - 9 | 689 | 62 | |
| GenBank mRNA | AK072434.1 | - | 6 - 7 | 2098 | 62 | |
| BT132393.1 | - | 8 - 9 | 629 | 62 | ||
More Information
Set Membership: |
|

LOC9270576
Genomic Map