Assay Details
| Gene Symbol: LOC4333161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774943.2 | XP_015630429.1 | 9 - 10 | 1917 | 82 | |
| GenBank mRNA | AK060637.1 | - | 5 - 6 | 878 | 82 | |
| AK069123.1 | - | 9 - 10 | 1924 | 82 | ||
More Information
Set Membership: |
|

LOC4333161
Genomic Map