Assay Details
| Gene Symbol: LOC4333522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773471.2 | XP_015628957.1 | 2 - 3 | 404 | 72 | |
| GenBank mRNA | AK073129.1 | - | 2 - 3 | 282 | 72 | |
| AK073903.1 | - | 2 - 3 | 405 | 72 | ||
More Information
Set Membership: |
|

LOC4333522
Genomic Map