Assay Details
| Gene Symbol: LOC4333604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772788.2 | XP_015628274.1 | 11 - 12 | 997 | 90 | |
| GenBank mRNA | AB255026.1 | - | 11 - 12 | 875 | 90 | |
| AK287869.1 | - | 11 - 12 | 998 | 90 | ||
More Information
Set Membership: |
|

LOC4333604
Genomic Map