Assay Details
Gene Symbol: LOC4334541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772583.2 | XP_015628069.1 | 1 - 2 | 289 | 120 | |
GenBank mRNA | AK103214.1 | - | 1 - 2 | 108 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772583.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
