Assay Details
Gene Symbol: LOC4334541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772583.2 | XP_015628069.1 | 6 - 7 | 1751 | 65 | |
GenBank mRNA | AK103214.1 | - | 6 - 7 | 1570 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
