Assay Details
| Gene Symbol: LOC4334609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777476.2 | XP_015632962.1 | 1 - 2 | 407 | 66 | |
| XM_015777477.2 | XP_015632963.1 | 1 - 2 | 407 | 66 | ||
| XM_015777478.2 | XP_015632964.1 | 1 - 2 | 407 | 66 | ||
| XM_015777479.2 | XP_015632965.1 | 1 - 2 | 200 | 66 | ||
| XM_015777480.2 | XP_015632966.1 | 1 - 2 | 200 | 66 | ||
| GenBank mRNA | AK060050.1 | - | 1 - 2 | 155 | 66 | |
More Information
Set Membership: |
|

LOC4334609
Genomic Map