Assay Details
| Gene Symbol: LOC4334651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774927.2 | XP_015630413.1 | 2 - 3 | 435 | 98 | |
| GenBank mRNA | AK059132.1 | - | 2 - 3 | 424 | 98 | |
| AK062799.1 | - | 2 - 3 | 395 | 98 | ||
| AK099043.1 | - | 2 - 3 | 419 | 98 | ||
More Information
Set Membership: |
|

LOC4334651
Genomic Map