Assay Details
Gene Symbol: LOC4334969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781152.2 | XP_015636638.1 | 2 - 3 | 938 | 75 | |
GenBank mRNA | AK061898.1 | - | 2 - 3 | 452 | 75 | |
AK065035.1 | - | 2 - 3 | 697 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
