Assay Details
Gene Symbol: LOC4334969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781152.2 | XP_015636638.1 | 4 - 5 | 1214 | 81 | |
GenBank mRNA | AJ698084.1 | - | 4 - 5 | 731 | 81 | |
AK061898.1 | - | 4 - 5 | 728 | 81 | ||
AK065035.1 | - | 4 - 5 | 973 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
