Assay Details
Gene Symbol: LOC4334981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778338.2 | XP_015633824.1 | 1 - 2 | 886 | 119 | |
GenBank mRNA | AK073177.1 | - | 1 - 2 | 880 | 119 | |
AK243157.1 | - | 1 - 2 | 906 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
