Assay Details
| Gene Symbol: LOC4335112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780494.2 | XP_015635980.1 | 5 - 6 | 2252 | 80 | |
| XM_015780496.2 | XP_015635982.1 | 4 - 5 | 2158 | 80 | ||
| GenBank mRNA | AK064939.1 | - | 5 - 6 | 2213 | 80 | |
More Information
Set Membership: |
|

LOC4335112
Genomic Map