Assay Details
Gene Symbol: LOC4335226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781216.1 | XP_015636702.1 | 3 - 4 | 908 | 77 | |
GenBank mRNA | AK061312.1 | - | 3 - 4 | 817 | 77 | |
AK061491.1 | - | 3 - 4 | 823 | 77 | ||
AK102155.1 | - | 3 - 4 | 829 | 77 | ||
AK119661.1 | - | 3 - 4 | 716 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
